missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Concentrate Na2CO3 72 mM in Water, IC eluent, Concentrate (20x) for Metrosep A Supp 7, MilliporeSigma™ Supelco™

Catalog No. 111014321
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014321 1 L
111014322 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014321 Supplier Merck Emd Millipore Supplier No. 727841L
Only null left

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Specifications

CAS 497-19-8
Quantity 1 L
Linear Formula Na2 CO3
MDL Number MFCD00003494
Synonym Sodium carbonate solution
Formula Weight 105.99
Concentration or Composition (by Analyte or Components) 72 mM Na2 CO3 aq. soln.