missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate concentrate, For 1L standard solution, 0.05Â M Na2CO3 (0.1N), Honeywell Fluka™
for 1L standard solution, 0.05Â M Na2CO3 (0.1N)
$480.92 - $480.92
Chemical Identifiers
| CAS | 497-19-8 |
|---|---|
| Molecular Formula | CNa2O3 |
| Molecular Weight (g/mol) | 105.99 |
| MDL Number | MFCD00003494 |
| InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
| Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
| PubChem CID | 10340 |
| ChEBI | CHEBI:29377 |
| IUPAC Name | disodium;carbonate |
| SMILES | [Na+].[Na+].[O-]C([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| disodium;carbonate |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 | |
| [Na+].[Na+].[O-]C([O-])=O |
Specifications
| 497-19-8 | |
| CNa2O3 | |
| MFCD00003494 | |
| 4154566 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium;carbonate | |
| 10340 | |
| 105.99g/mol | |
| Sodium carbonate concentrate |
| 1 Pc. | |
| Na2CO3 | |
| NONH for all modes of transport | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| Ampoule |
Safety and Handling
P305 + P351 + P338