missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Concentrate 0.1 M Na2CO3 in Water, Eluent concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111014324
Change view
Click to view available options
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111014324 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014324 Supplier Merck Emd Millipore Supplier No. 561691L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Prepared with sodium carbonate and high purity water (18.2 MOmega, 0.2 μm filtered)

Specifications

Chemical Name or Material Sodium Carbonate
CAS 497-19-8, 7732-18-5
Physical Form Liquid
Quantity 1 L
Linear Formula Na2CO3
MDL Number MFCD00003494
Synonym Sodium carbonate solution
Molecular Weight (g/mol) 105.99
Formula Weight 105.99
Packaging Poly Bottle