Learn More
Sodium Carbonate, Certified, 10.0% (w/v) ±0.5%, LabChem™
Supplier: LabChem LC229804
Specifications
| Sodium Carbonate | |
| Colorless | |
| Liquid | |
| CNa2O3 | |
| MFCD00003494 | |
| Soluble in water | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| 10.0% ±0.5% (w/v) | |
| Passes Test | |
| 1g/mL | |
| 1g/mL |
| 497-19-8,7732-18-5 | |
| 90,10 | |
| 4 L | |
| Na2CO3 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium carbonate | |
| 10340 | |
| 105.99 | |
| Certified | |
| Poly Bottle | |
| Fume; Carbon monoxide; Carbon dioxide |
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| disodium carbonate |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 | |
| [Na+].[Na+].[O-]C([O-])=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature