missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Anhydrous, Technical, Spectrum™ Chemical

Catalog No. 18613372
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613372 500 g Fiber Drum
18613369 100 lb. Fiber Drum
18613370 2.5 kg Fiber Drum
18613371 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613372 Supplier Spectrum Chemical Mfg Cor Supplier No. S1226500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 497-19-8

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Molecular Weight (g/mol) 105.99
Grade Technical
Packaging Fiber Drum
Chemical Name or Material Sodium Carbonate, Anhydrous

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.