missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Anhydrous (Powder/HPLC), Fisher Chemical

Catalog No. S495500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S495500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S495500 Supplier Thermo Fisher Scientific Supplier No. FLS495500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600°C
Melting Point 854°C
Color White
pH 11.3
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.5 %
Formula Weight 105.99
Percent Purity ≥99.5%
Grade Also meets ACS specifications
Identification Pass Test
Packaging Poly Bottle
Density 2.53g/cm³
Chemical Name or Material Sodium Carbonate Anhydrous
Show More Show Less