missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, 98%

Catalog No. p-7046087
Change view
Click to view available options
Quantity:
500 g
2500 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1309836 500 g
AAL130980E 2500 g
AAL130980I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1309836 Supplier Thermo Scientific Chemicals Supplier No. L1309836
Only null left

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol

Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.

Solubility
Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide, acetone, and alkyl acetates.

Notes
m.p. 851° (begins to lose CO{2} at 400°)It is hygroscopic in nature, and absorbs moisture from air. On heating, it begins to lose carbon dioxide at about 400 °C.

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

pH 11.5
Quantity 500 g
Beilstein 4154566
Merck Index 14,8596
Solubility Information Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates.
IUPAC Name disodium;carbonate
Formula Weight 105.99
Percent Purity 98%
Odor Odorless
Refractive Index 1.535
Sensitivity Hygroscopic
Density 2.53
Chemical Name or Material Sodium carbonate
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P234-P264b-P280i-P305+P351+P338-P390
EINECSNumber 207-838-8
RTECSNumber VZ4050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only