missing translation for 'onlineSavingsMsg'
Learn More

Sodium cacodylate, 0.1M buffer soln., pH 6.8, Thermo Scientific Chemicals

$265.90 - $265.90

Chemical Identifiers

CAS 6131-99-3
Molecular Weight (g/mol) 214.024
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62202AE
View Documents
Thermo Scientific Chemicals
J62202AE
100 mL
Each for $265.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6131-99-3
214.024

Specifications

6131-99-3
6.8
100 mL
0.1M buffer soln.
Liquid
214.024
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only