missing translation for 'onlineSavingsMsg'
Learn More

Sodium cacodylate, 0.1M buffer soln., pH 6.5

Catalog No. aaj60367ae
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60367AE 100 mL
AAJ60367AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAJ60367AE Supplier Thermo Scientific Chemicals Supplier No. J60367AE
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 6131-99-3214.024 g/mol

Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

Solubility
Soluble in water (∼100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 6131-99-3
Molecular Weight (g/mol) 214.024

Specifications

Concentration 0.1M buffer soln.
pH 6.5
Physical Form Liquid
Quantity 100 mL
Recommended Storage Keep cold

RUO – Research Use Only