missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromoacetate, 98%

Catalog No. aal1498314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1498314 25 g
AAL1498306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1498314 Supplier Thermo Scientific Chemicals Supplier No. L1498314

CAS: 1068-52-6 | C2H2BrNaO2 | 160.93 g/mol

It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1068-52-6
Molecular Formula C2H2BrNaO2
Molecular Weight (g/mol) 160.93
MDL Number MFCD01311832
InChI Key SESSOVUNEZQNBV-UHFFFAOYSA-M
Synonym sodium bromoacetate, sodium 2-bromoacetate, sodium monobromoacetate, unii-8j155ck8pv, acetic acid, bromo-, sodium salt, bromoacetic acid sodium salt, acmc-1bowb, sodium bromoacetate, ?brch2coona, acetic acid,bromo-,sodium salt 8ci,9ci, acetic acid, bromo-,sodium salt 8ci,9ci
PubChem CID 23675628
IUPAC Name sodium;2-bromoacetate
SMILES C(C(=O)[O-])Br.[Na+]

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Solubility Information Soluble in water.
Formula Weight 160.93
Percent Purity 98%
Chemical Name or Material Sodium bromoacetate
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
RTECSNumber AF6475000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only