missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, Honeywell Fluka™

Catalog No. p-7071696
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001802 500 g
6001800 1 kg
6001801 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001802 Supplier Honeywell Chemicals Supplier No. 02119500G
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Quantity 500 g
Linear Formula NaBr
UN Number NONH for all modes of transport
IUPAC Name sodium bromide
Formula Weight 102.89g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 1mmHg (806°C)
Chemical Name or Material Sodium Bromide, Meets analytical specification of Ph. Eur.BP,USP
EINECSNumber 231-599-9
RTECSNumber VZ3150000