missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bromide, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | NaBr |
| UN Number | NONH for all modes of transport |
| IUPAC Name | sodium bromide |
| Formula Weight | 102.89g/mol |
| Percent Purity | 99 to 100.5% (calc. to the dried substance) |
| Grade | puriss. |
| Packaging | Plastic Bottle |
| Vapor Pressure | 1mmHg (806°C) |
| Chemical Name or Material | Sodium Bromide, Meets analytical specification of Ph. Eur.BP,USP |
Safety and Handling
| EINECSNumber | 231-599-9 |
| RTECSNumber | VZ3150000 |