missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide (Granular/Certified ACS), Fisher Chemical™

Catalog No. p-33675
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S255500 500 g Glass Bottle
S2553 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S255500 Supplier Thermo Fisher Scientific Supplier No. S255 500
Only null left

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Color White
pH 5 to 8.8
Physical Form Powder
Quantity 500 g
Assay Percent Range ≥99 %
Formula Weight 102.89
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Sodium Bromide
Show More Show Less