missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, Granular, ACS, 99%, Spectrum™ Chemical

Catalog No. 18613333
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613333 2.5 kg Amber Glass Bottle
18613329 125 g Amber Glass Bottle
18613331 500 g Amber Glass Bottle
18613330 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613333 Supplier Spectrum Chemical Mfg Cor Supplier No. S11952.5KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

CAS Max % 1
pH 5.0 to 8.8
Physical Form Granular/Lump
Quantity 2.5 kg
Percent Purity 99%
Grade ACS/Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Bromide, Granular

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.