missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390884
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004008 500 g Poly Bottle
02004007 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004008 Supplier Avantor Supplier No. 358801
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Physical Form Crystals
Quantity 500 g
IUPAC Name sodium;bromide
Formula Weight 102.89
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.21kg
Chemical Name or Material Sodium Bromide