missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bromide, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1390.0°C |
| Melting Point | 755.0°C |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Packaging | Glass bottle |
| Linear Formula | NaBr |
| Fieser | 12,445; 13,46 |
| Merck Index | 15,8729 |
| Solubility Information | Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol |
| IUPAC Name | sodium bromide |
| Show More |
Safety and Handling
| EINECSNumber | 231-599-9 |
RUO – Research Use Only