missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. lc229611
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC229611 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC229611 Supplier LabChem Supplier No. LC229611

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1,390°C
Melting Point 755°C
Color White
CAS Max % 100
Physical Form Crystalline
Quantity 500 g
Linear Formula NaBr
Solubility Information Soluble in water
Formula Weight 102.89
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen bromide, Bromine
Chemical Name or Material Sodium Bromide
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 231-599-9
Recommended Storage Room Temperature