missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, ACS, 99.0% min

Catalog No. AA3644436
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3644436 500 g
AA36444A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3644436 Supplier Thermo Scientific Chemicals Supplier No. 03644436

CAS: 7647-15-6 | BrNa | 102.89 g/mol

It is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0%
Merck Index 14,8594
Solubility Information Soluble in water,soluble in alcohol.
IUPAC Name sodium;bromide
Formula Weight 102.89
Percent Purity ≤99.0%
Grade ACS
Refractive Index 1.6412
Sensitivity Hygroscopic
Density 3.203 g/mL
Chemical Name or Material Sodium bromide, For analysis ACS
Show More Show Less
EINECSNumber 231-599-9
RTECSNumber VZ3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only