missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99+%, extra pure, anhydrous

Catalog No. p-3735075
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205131000 100 g Plastic bottle
AC205130010 1 kg Plastic bottle
AC205130050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205131000 Supplier Thermo Scientific Chemicals Supplier No. 205131000
Only null left

CAS: 7647-15-6 | BrNa | 102.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390.0°C
Melting Point 755.0°C
Color White
pH 5 to 7 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15, 8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
Formula Weight 102.89
Percent Purity ≥99%
Grade Extra Pure
Loss on Drying 1% max. (120°C)
Packaging Plastic bottle
Chemical Name or Material Sodium bromide, Anhydrous
Show More Show Less
EINECSNumber 231-599-9

RUO – Research Use Only