missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99.99% (metals basis)

Catalog No. p-4782137
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8760406 5 g
AA8760414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8760406 Supplier Thermo Scientific Chemicals Supplier No. 08760406
Only null left

CAS: 7647-15-6 | BrNa | 102.89 g/mol

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water and alcohol.
IUPAC Name sodium bromide
Formula Weight 102.89
Refractive Index 1.6412
Sensitivity Hygroscopic
Density 3.203 g/cm3 at 20°C
Chemical Name or Material Sodium bromide
Show More Show Less
EINECSNumber 231-599-9
RTECSNumber VZ3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only