missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bromide, 99.5%, for analysis
CAS: 7647-15-6 | BrNa | 102.89 g/mol
Supplier: Thermo Scientific Chemicals 246900025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
| 1390.0°C | |
| 755.0°C | |
| 7647-15-6 | |
| 5 to 8.8 (5% soln. at 20°C) | |
| 2.5 kg | |
| BrNa | |
| MFCD00003475 | |
| 15, 8729 | |
| Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C),1210g/L water (100°C),1g/6 mL methanol,7g/100 mL 95% ethanol | |
| [Na+].[Br-] | |
| 102.89 | |
| CHEBI:63004 | |
| 99.5% | |
| Plastic bottle |
| Sodium bromide | |
| p.a. | |
| White | |
| Crystalline Powder | |
| 99.5% min. (Argentometry) | |
| NaBr | |
| 12,445; 13,46 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| sodium bromide | |
| 253881 | |
| 102.89 | |
| Analytical |
Chemical Identifiers
| 7647-15-6 | |
| 102.89 | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| 253881 | |
| [Na+].[Br-] |
| BrNa | |
| MFCD00003475 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| CHEBI:63004 |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 231-599-9
RTECSNumber : VZ3150000
TSCA : TSCA
RUO – Research Use Only