missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, 99.0% min., ACS, MilliporeSigma™

Catalog No. MSX03901
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX03901 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX03901 Supplier Technidata Supplier No. SX03901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity sodium bromide reagent that meets specifications for analytical and pharmaceutical applications.

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0% min.
Linear Formula NaBr
Formula Weight 12.89
Percent Purity ≥99.0%
Grade ACS
Vapor Pressure 1.3 hPa (806°C)
Density 3.20g/cm3 (20°C); Bulk Density: 1750 kg/m3
Chemical Name or Material Sodium Bromide, Suprapur™
Show More Show Less
Recommended Storage Store without restrictions