missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bromide, ≥99.0%, ACS Reagent, Honeywell™ Fluka™
ACS Reagent,≥99.0%
$307.08 - $307.08
Chemical Identifiers
| CAS | 7647-15-6 |
|---|---|
| Molecular Formula | BrNa |
| Molecular Weight (g/mol) | 102.89 |
| MDL Number | MFCD00003475 |
| InChI Key | JHJLBTNAGRQEKS-UHFFFAOYSA-M |
| Synonym | sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a |
| PubChem CID | 253881 |
| ChEBI | CHEBI:63004 |
| IUPAC Name | sodium;bromide |
| SMILES | [Na+].[Br-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-15-6 | |
| 102.89 | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| 253881 | |
| sodium;bromide |
| BrNa | |
| MFCD00003475 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| CHEBI:63004 | |
| [Na+].[Br-] |
Specifications
| 1393°C | |
| 7647-15-6 | |
| BrNa | |
| MFCD00003475 | |
| sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
| JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
| sodium;bromide | |
| 253881 | |
| 102.89g/mol | |
| ACS Reagent | |
| 1mmHg (806°C) | |
| Sodium Bromide |
| 755°C | |
| 500 g | |
| NaBr | |
| NONH for all modes of transport | |
| Soluble in water and ethanol | |
| [Na+].[Br-] | |
| 102.89 | |
| CHEBI:63004 | |
| ≥99.0% | |
| Poly Bottle | |
| 3.2g/cm3 |
Safety and Handling
EINECSNumber : 231-599-9
RTECSNumber : VZ3150000