missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium borohydride, (VenPureTM solution), 12% aq. solution
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 130.0°C to 135.0°C |
| Chemical Name or Material | Sodium borohydride |
| Name Note | VenPure™ solution, 12% aq. solution |
| CAS | 16940-66-2, 1310-73-2, 67-56-1, 7732-18-5 |
| Quantity | 100 mL |
| Molecular Formula | BH4Na |
| Linear Formula | NaBH4 |
| MDL Number | MFCD00003518 |
| Merck Index | 15,8727 |
| Synonym | SBH, Sodium tetrahydroborate |
| Show More |
RUO – Research Use Only