missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

Supplier:  Thermo Scientific Chemicals 429131000

Catalog No. AC429131000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

130.0°C to 135.0°C
16940-66-2,1310-73-2,67-56-1,7732-18-5
Liquid
BH4Na
MFCD00003518
Sodium tetrahydroborate, SBH
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.82
AcroSeal™ Glass Bottle
1.4000g/mL
Sodium borohydride
Colorless
100 mL
NaBH4
15,8727
Solubility in water: completely soluble
[BH4-].[Na+]
37.83
11 to 13%
1.4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only