missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, Certified, 40g/L, LabChem™

Catalog No. LC229601
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC229601 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC229601 Supplier LabChem Supplier No. LC229601

Specifications

Chemical Name or Material Sodium Borate
Color Colorless
CAS 7732-18-5, 1303-96-4, 1310-73-2
pH 9.5 ±0.1
CAS Max % 95.98, 4, 0.02
Physical Form Liquid
Quantity 500 mL
Linear Formula Na2B4O7·10H2O
Synonym Borax; Sodium Tetraborate
Solubility Information Soluble in water
Formula Weight 381.37
Grade Certified
Concentration or Composition (by Analyte or Components) 40g/L
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Boron
Density 1g/mL
Show More Show Less