missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate Buffer, Ricca Chemical

Catalog No. R71499001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R71499001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R71499001A Supplier Ricca Chemical Company Supplier No. R71499001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

TAPPI for Specific External Surface of Pulp

Specifications

Color Colorless
Quantity 1 L
Packaging Natural Poly Bottle
CAS 7732-18-5
Form Liquid
Buffer Yes