missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Borate Buffer, pH 9.9, for Sulfite Analysis, Reagents
Specifications
Specifications
| Chemical Name or Material | Buffer |
| CAS | 7732-18-5 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Sterility | Non-sterile |
| Solubility Information | Completely miscible |