missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bisulfite, Purified *This material is a mixture containing some Sodium Bisulfite (CAS# 7631-90-5), Reagents
Sodium Bisulfite, Purified *This material is a mixture containing some Sodium Bisulfite (CAS# 7631-90-5), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
$192.23 - $980.20
Chemical Identifiers
| CAS | 7681-57-4 |
|---|---|
| Molecular Formula | Na2O5S2 |
| Molecular Weight (g/mol) | 190.09 |
| InChI Key | LDTLADDKFLAYJA-UHFFFAOYSA-L |
| Synonym | Sodium Hydrogen Sulfite |
| IUPAC Name | disodium (sulfinatooxy)sulfinate |
| SMILES | [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-055
|
Labchem Inc
C231000025A5 |
2.5 kg |
Each for $576.83
|
|
|||||
|
01-337-054
|
Labchem Inc
C2310000500A5 |
500 g |
Each for $192.23
|
|
|||||
|
01-337-053
|
Labchem Inc
C231000050F5 |
50 lb. |
Each for $980.20
|
|
|||||
Chemical Identifiers
| 7681-57-4 | |
| 190.09 | |
| Sodium Hydrogen Sulfite | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Na2O5S2 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| disodium (sulfinatooxy)sulfinate |
Specifications
| 315°C | |
| Colorless/White | |
| Solid | |
| Na2O5S2 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| disodium (sulfinatooxy)sulfinate | |
| Pure | |
| 1.48 |
| 150°C | |
| 7681-57-4 | |
| 2.5 kg | |
| Sodium Hydrogen Sulfite | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O | |
| 190.09 | |
| Poly Bottle | |
| Sodium Bisulfite, Purified *This material is a mixture containing some Sodium Bisulfite (CAS# 7631-90-5) |