missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, Purified *This material is a mixture containing some Sodium Bisulfite (CAS# 7631-90-5), Reagents

Catalog No. 01337055
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337055 2.5 kg
01337054 500 g
01337053 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337055 Supplier Labchem Inc Supplier No. C23100002.5A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Bisulfite, Purified *This material is a mixture containing some Sodium Bisulfite (CAS# 7631-90-5), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym Sodium Hydrogen Sulfite
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Boiling Point 315°C
Melting Point 150°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.48
Chemical Name or Material Sodium Bisulfite, Purified *This material is a mixture containing some Sodium Bisulfite (CAS# 7631-90-5)