missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, Granular, FCC, 58.5-67.4%, Spectrum™ Chemical

Catalog No. p-7158187
Change view
Click to view available options
Quantity:
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606342 2.5 kg Poly Bottle
18606344 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606342 Supplier Spectrum Chemical Mfg Cor Supplier No. S11722.5KG

S1172, 7631-90-5, NaHSO3

Spectrum™ Chemical Sodium Bisulfite, Granular, FCC is used as an antioxidant to preserve the aromas and flavors. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 58.5 to 67.4%
Grade FCC
Packaging Poly Bottle