missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite, Granular, FCC, 58.5-67.4%, Spectrum™ Chemical

Catalog No. p-7158503
Change view
Click to view available options
Quantity:
500 g
12 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606341 500 g Poly Bottle
18606343 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606341 Supplier Spectrum Chemical Mfg Cor Supplier No. S1172500GM

S1172, 7631-90-5, NaHSO3

Spectrum™ Chemical Sodium Bisulfite, Granular, FCC is used as an antioxidant to preserve the aromas and flavors. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 58.5 to 67.4%
Grade FCC
Packaging Poly Bottle