missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfate, Solstice

Catalog No. 6002849
Change view
Click to view available options
Quantity:
250 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002849 5 kg
6002848 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002849 Supplier Solstice Supplier No. 716565KG
Only null left

Purum, anhydrous, ∼95.0%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
IUPAC Name sodium;hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O

Specifications

Quantity 5 kg
Linear Formula NaHSO4
UN Number NONH for all modes of transport
Formula Weight 120.06g/mol
Percent Purity 93 to 102% (T), 95%
Chemical Name or Material Sodium bisulfate
Hazard Statement P280-P305 + P351 + P338
RTECSNumber VZ1860000