missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bisulfate, Honeywell Fluka™
Purum, anhydrous, ∼95.0%
Supplier: Honeywell Chemicals 716561KG
Description
- ≥95% for general laboratory use.
Specifications
| Sodium bisulfate | |
| 1 kg | |
| NaHSO4 | |
| NONH for all modes of transport | |
| WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
| sodium;hydrogen sulfate | |
| 516919 | |
| 93 to 102% (T), 95% |
| 7681-38-1 | |
| HNaO4S | |
| MFCD00003531 | |
| sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
| [Na+].OS([O-])(=O)=O | |
| 120.05 | |
| 120.06g/mol |
Chemical Identifiers
| 7681-38-1 | |
| 120.05 | |
| WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
| 516919 | |
| [Na+].OS([O-])(=O)=O |
| HNaO4S | |
| MFCD00003531 | |
| sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
| sodium;hydrogen sulfate |
Safety and Handling
P280-P305 + P351 + P338
RTECSNumber : VZ1860000