missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfate, Anhydrous, Reagents

Catalog No. p-200021179
Change view
Click to view available options
Quantity:
100 g
12 kg
500 g
Catalog No. Quantity
01337058 100 g
01337057 12 kg
01337056 500 g
3 options

Catalog No. 01337058

Supplier: Labchem Inc C2309500100C2

Only null left

Sodium Bisulfate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym Sodium hydrogen sulfate
IUPAC Name sodium hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.4394
Chemical Name or Material Sodium Bisulfate, Anhydrous