missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfate, 92%, technical

Catalog No. p-3777063
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC214850250 25 g Glass bottle
AC214850010 1 kg Plastic Bottle
AC214850025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC214850250 Supplier Thermo Scientific Chemicals Supplier No. 214850250
Only null left

CAS: 7681-38-1 | HNaO4S | 120.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
IUPAC Name sodium hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O

Specifications

Melting Point 177.0°C to 180.0°C
Color White to Yellow
Physical Form Granules
Quantity 25 g
Assay Percent Range 92%
Linear Formula NaHSO4
Merck Index 15, 8722
Solubility Information Solubility in water: soluble. Other solubilities: decomposes by alcohol into na2so4 and h2so4
Formula Weight 120.06
Percent Purity 92%
Grade Technical
Packaging Glass bottle
Specific Gravity 2.1
Density 2.1000g/mL
Chemical Name or Material Sodium bisulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 231-665-7

RUO – Research Use Only