missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. S09895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09895G 5 g
S09891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09895G Supplier TCI America Supplier No. S09895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91742-21-1
Molecular Formula C2HF6NNaO4S2
Molecular Weight (g/mol) 304.133
MDL Number MFCD14156021
InChI Key QXZNUMVOKMLCEX-UHFFFAOYSA-N
Synonym Bis(trifluoromethanesulfonyl)imide Sodium Salt, Sodium Triflimide
PubChem CID 131887538
IUPAC Name sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]

Specifications

Melting Point 257°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 303.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Bis(trifluoromethanesulfonyl)imide