missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Catalog No. S09921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09921G 1 g
S09925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09921G Supplier TCI America Supplier No. S09921G
Only null left

Chemical Identifiers

CAS 100669-96-3
Molecular Formula F2NNaO4S2
Molecular Weight (g/mol) 203.11
InChI Key VCCATSJUUVERFU-UHFFFAOYSA-N
PubChem CID 102310776
IUPAC Name sodium;bis(fluorosulfonyl)azanide
SMILES [N-](S(=O)(=O)F)S(=O)(=O)F.[Na+]

Specifications

Melting Point 112°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 203.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Bis(fluorosulfonyl)imide