missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bifluoride, Technical, Spectrum™ Chemical

$337.93 - $6107.30

Chemical Identifiers

CAS 1333-83-1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613303
View Documents
Spectrum Chemical Mfg Cor
S1160500GM
500 g Poly Bottle
Each for $337.93
Only null left
 
18613301
View Documents
Spectrum Chemical Mfg Cor
S116025KG
2.5 kg Poly Bottle
Each for $1,106.97
Only null left
 
18613302
View Documents
Spectrum Chemical Mfg Cor
S116012KG
12 kg Poly Pail
Each for $2,744.60
Only null left
 
18613304
View Documents
Spectrum Chemical Mfg Cor
S116050KGBL
50 kg Fiber Drum
Each for $6,107.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1333-83-1

Specifications

1333-83-1
500 g
Poly Bottle
1
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.