Learn More
Sodium beta-glycerophosphate pentahydrate, 97%
CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol
Supplier: Thermo Scientific Chemicals L0342522
Description
Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
| Sodium beta-glycerophosphate pentahydrate | |
| >300°C | |
| C3H19Na2O11P | |
| 100 g | |
| 14,8622 | |
| Soluble in water. | |
| O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O | |
| 308.13 | |
| 306.12 (216.04 Anhydrous) |
| 13408-09-8 | |
| 9.5 | |
| MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 | |
| 3744328 | |
| unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate | |
| VWVLGHQKEJESPW-UHFFFAOYSA-N | |
| 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate | |
| 131846916 | |
| 97% |
Chemical Identifiers
| 13408-09-8 | |
| 308.13 | |
| VWVLGHQKEJESPW-UHFFFAOYSA-N | |
| 131846916 | |
| O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O |
| C3H19Na2O11P | |
| MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 | |
| unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate | |
| 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate |
Safety and Handling
EINECSNumber : 212-464-3
RTECSNumber : UA0600000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only