missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Benzoate, Technical, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Crystalline Powder or Needles |
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Packaging | Poly Pail |
| Molecular Weight (g/mol) | 144.11 |
| Grade | Technical |
| Chemical Name or Material | Sodium Benzoate |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.