missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Purified, Reagents

Catalog No. 01337063
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337063 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337063 Supplier Labchem Inc Supplier No. C2308000500A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Benzoate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
IUPAC Name sodium benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Boiling Range >450°C
Melting Point 410°C to 430°C
Color White to Off-White
Boiling Point >450°C
Melting Range 410°C to 430°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1.44
Grade Pure
Physical Form Solid
Chemical Name or Material Sodium Benzoate, Purified
Show More Show Less