missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Powder, FCC, 99-101%, Spectrum™ Chemical

Catalog No. 18606313
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606313 500 g Poly Bottle
18606315 2.5 kg Poly Pail
18606314 6 x 500 g Poly Bottle
18606316 4 x 2.5 kg Poly Pail
18606317 12 kg Poly Pail
18606318 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606313 Supplier Spectrum Chemical Mfg Cor Supplier No. S1146500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1146, 532-32-1, C7H5O2Na

Spectrum™ Chemical Sodium Benzoate, Powder, FCC is a widely used food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
IUPAC Name sodium benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade FCC