missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Powder, 99%, Spectrum™ Chemical

Catalog No. p-7166287
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18613286 500 g Poly Bottle
18613287 2.5 kg Poly Pail
2 options

Catalog No. 18613286

Supplier: Spectrum Chemical Mfg Cor S1143500GM

Only null left

Chemical Identifiers

CAS 532-32-1

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.