missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Powder, 99%, Spectrum™ Chemical

Catalog No. 18613286
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613286 500 g Poly Bottle
18613287 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18613286 Supplier Spectrum Chemical Mfg Cor Supplier No. S1143500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 532-32-1

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.