missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate 98.0+%, TCI America™

Supplier:  TCI America S085525G

 View more versions of this product

Catalog No. S085525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Benzoate [for Biochemical Research]
450°C
C7H5NaO2
25 g
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate
517055
≥98.0% (HPLC,T)
532-32-1
White
MFCD00012463
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
[Na+].[O-]C(=O)C1=CC=CC=C1
144.11
144.10
Crystalline Powder

Chemical Identifiers

532-32-1
144.11
WXMKPNITSTVMEF-UHFFFAOYSA-M
517055
[Na+].[O-]C(=O)C1=CC=CC=C1
C7H5NaO2
MFCD00012463
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
sodium benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1272&(3)-1293

RTECSNumber : DH6650000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions