missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate 98.0+%, TCI America™

Catalog No. S08555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08555G 5 g
S085525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08555G Supplier TCI America Supplier No. S08555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
IUPAC Name sodium benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point 450°C
Color White
Quantity 5 g
Formula Weight 144.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Benzoate [for Biochemical Research]
EINECSNumber (3)-1272&(3)-1293
RTECSNumber DH6650000
TSCA Yes