missing translation for 'onlineSavingsMsg'
Learn More

Sodium benzoate, 99%

Catalog No. AAA1594636
Change view
Click to view available options
Quantity:
10,000 g
250 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1594636 500 g
AAA1594630 250 g
AAA159460E 2500 g
AAA159460C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1594636 Supplier Thermo Scientific Chemicals Supplier No. A1594636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium benzoate is widely used as a preservative in food, medicine, cosmetics and animal feeds. It is used in the treatment of hyperammonemia and urea cycle disorders. It is used in the fireworks as a fuel in whistle mix. It is also used in the preparation of toothpaste and mouthwashes. It finds application in most of the acidic foods such as salad dressings (vinegar), carbonated drinks (carbonic acid), jams and fruit juices (citric acid), pickles (vinegar), and condiments.

Solubility
Soluble in water, alcoholSoluble in water, alcohol, ammonia and pyridine.

Notes
Hygroscopic. Incompatible with alkalis, mineral acids and strong oxidizing agents.

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 500 g
Beilstein 3572467
Merck Index 14,8582
Solubility Information Soluble in water,alcoholSoluble in water,alcohol,ammonia and pyridine.
IUPAC Name sodium benzoate
Formula Weight 144.11
Percent Purity 99%
Physical Form Powder or granules or prills
Chemical Name or Material Sodium Benzoate
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 208-534-8
RTECSNumber DH6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only