missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzenesulfonate 96.0+%, TCI America™

Catalog No. p-7118314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
B003525G 25 g
B0035100G 100 g
B0035500G 500 g
3 options

Catalog No. B003525G

Supplier: TCI America B003525G

Only null left

Chemical Identifiers

CAS 515-42-4
Molecular Formula C6H5NaO3S
Molecular Weight (g/mol) 180.153
MDL Number MFCD00065179
InChI Key MZSDGDXXBZSFTG-UHFFFAOYSA-M
Synonym sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate
PubChem CID 517327
IUPAC Name sodium;benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 180.15
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Benzenesulfonate
EINECSNumber (3)-1928
RTECSNumber DB7350000
TSCA Yes