missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzenesulfinate Dihydrate 98.0+%, TCI America™

Catalog No. p-7118308
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B002725G 25 g
B0027100G 100 g
B0027500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B002725G Supplier TCI America Supplier No. B002725G

Chemical Identifiers

CAS 25932-11-0
Molecular Formula C6H9NaO4S
Molecular Weight (g/mol) 200.184
MDL Number MFCD00149638
InChI Key MYXJYAIKMQJHIB-UHFFFAOYSA-M
Synonym sodium benzenesulfinate dihydrate, benzenesulfinic acid, sodium salt, dihydrate, sodium phenyl sulfinate dihydrate, benzensulfinan sodny czech, benzenesulfinic acid sodium salt dihydrate, benzensulfinan sodny, sodium benzenesulfinate hydrate 1:1:2, acmc-209gnh, phso2na.2h2o, ksc556o7r
PubChem CID 23681562
IUPAC Name sodium;benzenesulfinate;dihydrate
SMILES C1=CC=C(C=C1)S(=O)[O-].O.O.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 164.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Benzenesulfinate Dihydrate
EINECSNumber (3)-3123
RTECSNumber DA9135000
TSCA Yes